Merck
  • Flavonoids and Other Phenolic Compounds in Needles of Pinus peuce and Other Pine Species from the Macedonian Flora.
Flavonoids and Other Phenolic Compounds in Needles of Pinus peuce and Other Pine Species from the Macedonian Flora.
Natural product communications (2015-07-23)
Marija Karapandzova, Gjose Stefkov, Ivana Cvetkovikj, Jasmina Petreska Stanoeva, Marina Stefova, Svetlana Kulevanova
ABSTRACT
Flavonoids and other phenolic compounds in young needles of four pine species, Pinus peuce, P. nigra, P. mugo and P. sylvestris from the Macedonian flora were investigated. The amount of total phenols and total flavonoids were determined using Folin-Ciocalteau and aluminum chloride assay, respectively. The obtained results revealed that the total phenolic content (TPC) and total flavonoids content (TFC) varied among different pine species ranging from 9.8 to 14.0 mg GAE/g and from 3.3 to 7.2 mg CE/g of dried plant material, respectively. Qualitative analysis of flavonoids and other phenolic components was made by a LC-DAD/ESI-MS(n) optimized chromatographic method. A total of 17 phenolic components were identified and classified as: acids (2), procyanidins (2) and flavonoid glycosides (13). The most prevalent components were flavonoid glycosides, especially flavonols and methylated flavonols (9). Additionally, 3 components were found as acylated flavonol glycosides with ferulic and p-coumaric acid. The last one was found not only in esterified form but also in the free form. Only one flavone-apigenin glycoside was detected. Procyanidins were identified as catechin derivatives, both dimers and trimers.
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Aluminum chloride, SAJ first grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium carbonate, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium carbonate, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 20 % (w/v)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 7 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 0.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 0.02 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium hydroxide solution, 4 M