Merck
  • Passive sampling: A cost-effective method for understanding antibiotic fate, behaviour and impact.
Passive sampling: A cost-effective method for understanding antibiotic fate, behaviour and impact.
Environment international (2015-10-10)
Chang-Er Chen, Hao Zhang, Guang-Guo Ying, Li-Jun Zhou, Kevin C Jones
ABSTRACT
The occurrence of antibiotics in the environment has raised much concern in recent years. Understanding their release, fate and behaviour in the environment is vital to assess potential risks. In this study, a novel passive water sampler - diffusion gradients in thin-films for organics (o-DGT) - was employed to assess the occurrence and removal of antibiotics in two waste water treatment plants (WWTPs) - one in China and the other in the United Kingdom (UK). Of the targeted compounds, 11 of 19 were detected in the Chinese WWTP (ND-200ng/L) and 10 of 40 were found in the UK plant (ND-1380ng/L). Florfenicol, lincomycin, ofloxacin and roxithromycin were most abundant in the Chinese WWTP (influent), while anhydrous erythromycin, ciprofloxacin, trimethoprim, ofloxacin and sulfapyridine were the most abundant in the UK influent samples. Estimated Chinese and UK consumption data are used to interpret the results. Neither of the WWTPs was very effective at removing antibiotics: ~40-50% (overall) was removed by the two plants, with the rest being discharged into the receiving rivers. This is the first study using o-DGT to assess the occurrence and removal of antibiotics in WWTPs. o-DGT is a useful, cost-effective tool to assess WWTP performance and can highlight the effectiveness of treatment steps, which can be applied to wastewater based epidemiology studies.
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, SAJ first grade, ≥97.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, JIS special grade, ≥97.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), suitable for iron determination
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sulfanilamide, JIS special grade, ≥99.7%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), for copper determination
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sulfanilamide, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sulfamonomethoxine
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sulfaguanidine
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Salinomycin, from Streptomyces albus, ≥98% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sulfameter
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Trimethoprim, ≥98.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available. Contact Technical Service