Merck
  • Biochemical conversion of sugarcane straw hemicellulosic hydrolyzate supplemented with co-substrates for xylitol production.
Biochemical conversion of sugarcane straw hemicellulosic hydrolyzate supplemented with co-substrates for xylitol production.
Bioresource technology (2015-12-01)
A F Hernández-Pérez, I A L Costa, D D V Silva, K J Dussán, T R Villela, E V Canettieri, J A Carvalho, T G Soares Neto, M G A Felipe
ABSTRACT
Biotechnological production of xylitol is an attractive route to add value to a sugarcane biorefinery, through utilization of the hemicellulosic fraction of sugarcane straw, whose availability is increasing in Brazil. Herein, supplementation of the sugarcane straw hemicellulosic hydrolyzate (xylose 57gL(-1)) with maltose, sucrose, cellobiose or glycerol was proposed, and their effect as co-substrates on xylitol production by Candida guilliermondii FTI 20037 was studied. Sucrose (10gL(-1)) and glycerol (0.7gL(-1)) supplementation led to significant increase of 8.88% and 6.86% on xylose uptake rate (1.11gL(-1)h(-1) and 1.09gL(-1)), respectively, but only with sucrose, significant increments of 12.88% and 8.69% on final xylitol concentration (36.11gL(-1)) and volumetric productivity (0.75gL(-1)h(-1)), respectively, were achieved. Based on these results, utilization of complex sources of sucrose, derived from agro-industries, as nutritional supplementation for xylitol production can be proposed as a strategy for improving the yeast performance and reducing the cost of this bioprocess by replacing more expensive nutrients.
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Xylose, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerin, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)