Merck
Regulation of the anaphase-promoting complex by the COP9 signalosome.
Cell cycle (Georgetown, Tex.) (2009-06-19)
Robert Kob, Juliane Kelm, Nicole Posorski, Aria Baniahmad, Ferdinand von Eggeling, Christian Melle
ABSTRACT
The COP9 complex (signalosome) is a known regulator of the proteasome/ubiquitin pathway. Furthermore it regulates the activity of the cullin-RING ligase (CRL) families of ubiquitin E3-complexes. Besides the CRL family, the anaphase-promoting complex (APC/C) is a major regulator of the cell cycle. To investigate a possible connection between both complexes we assessed interacting partners of COP9 using an in vivo protein-protein interaction assay. Hereby, we were able to show for the first time that CSN2, a subunit of the COP9 signalosome, interacts physically with APC/C. Furthermore, we detected a functional influence of the COP9 complex regarding the stability of several targets of the APC/C. Consistent with these data we showed a genetic instability of cells overexpressing CSN2.