Merck
  • Sequencing chromosomal abnormalities reveals neurodevelopmental loci that confer risk across diagnostic boundaries.
Sequencing chromosomal abnormalities reveals neurodevelopmental loci that confer risk across diagnostic boundaries.
Cell (2012-04-24)
Michael E Talkowski, Jill A Rosenfeld, Ian Blumenthal, Vamsee Pillalamarri, Colby Chiang, Adrian Heilbut, Carl Ernst, Carrie Hanscom, Elizabeth Rossin, Amelia M Lindgren, Shahrin Pereira, Douglas Ruderfer, Andrew Kirby, Stephan Ripke, David J Harris, Ji-Hyun Lee, Kyungsoo Ha, Hyung-Goo Kim, Benjamin D Solomon, Andrea L Gropman, Diane Lucente, Katherine Sims, Toshiro K Ohsumi, Mark L Borowsky, Stephanie Loranger, Bradley Quade, Kasper Lage, Judith Miles, Bai-Lin Wu, Yiping Shen, Benjamin Neale, Lisa G Shaffer, Mark J Daly, Cynthia C Morton, James F Gusella
ABSTRACT
Balanced chromosomal abnormalities (BCAs) represent a relatively untapped reservoir of single-gene disruptions in neurodevelopmental disorders (NDDs). We sequenced BCAs in patients with autism or related NDDs, revealing disruption of 33 loci in four general categories: (1) genes previously associated with abnormal neurodevelopment (e.g., AUTS2, FOXP1, and CDKL5), (2) single-gene contributors to microdeletion syndromes (MBD5, SATB2, EHMT1, and SNURF-SNRPN), (3) novel risk loci (e.g., CHD8, KIRREL3, and ZNF507), and (4) genes associated with later-onset psychiatric disorders (e.g., TCF4, ZNF804A, PDE10A, GRIN2B, and ANK3). We also discovered among neurodevelopmental cases a profoundly increased burden of copy-number variants from these 33 loci and a significant enrichment of polygenic risk alleles from genome-wide association studies of autism and schizophrenia. Our findings suggest a polygenic risk model of autism and reveal that some neurodevelopmental genes are sensitive to perturbation by multiple mutational mechanisms, leading to variable phenotypic outcomes that manifest at different life stages.