Merck
  • Me3(OMe)tBuXPhos: a surrogate ligand for Me4tBuXPhos in palladium-catalyzed C-N and C-O bond-forming reactions.
Me3(OMe)tBuXPhos: a surrogate ligand for Me4tBuXPhos in palladium-catalyzed C-N and C-O bond-forming reactions.
The Journal of organic chemistry (2012-02-09)
Satoshi Ueda, Siraj Ali, Brett P Fors, Stephen L Buchwald
ABSTRACT
A new biarylphosphine ligand, Me(3)(OMe)tBuXPhos (L3), was designed as a surrogate for Me(4)tBuXPhos (L1). The Me(3)(OMe)tBuXPhos could be prepared in a chromatography-free manner from inexpensive and readily available 2,3,6-trimethylphenol. Comparative studies demonstrated that a catalyst based on Me(3)(OMe)tBuXPhos displayed the same reactivity as a catalyst based on Me(4)tBuXPhos for Pd-catalyzed C-N and C-O bond-forming processes.
Sigma-Aldrich
Me3(OMe)tBuXPhos, 96% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
3-Amino-5-bromobenzotrifluoride, 97%
Pricing and availability is not currently available.