Merck
  • New therapeutic schedule for prostatic cancer-3 cells with ET-1 RNAi and Endostar.
New therapeutic schedule for prostatic cancer-3 cells with ET-1 RNAi and Endostar.
Asian Pacific journal of cancer prevention : APJCP (2015-01-06)
Hao-Jie Zhang, Wei-Qing Qian, Ran Chen, Zhong-Quan Sun, Jian-Da Song, Lu Sheng
ABSTRACT
Endothelin-1 and Endostar are both significant for the progression, proliferation, metastasis and invasion of cancer. In this paper, we studied the effect of ET-1 RNAi and Endostar in PC-3 prostatic cancer cells. The lentiviral vector was used in the establishment of ET-1 knockdown PC-3 cells. Progression and apoptosis were assessed by CKK-8 and flow cytometry, respectively. Transwell assay was used to estimate invasion and signaling pathways were studied by Western blotting. ET-1 mRNA and protein in ET-1 knockdown PC-3 cells were reduced to 26.4% and 22.4% compared with control group, respectively. ET-1 RNAi and Endostar both were effective for the suppression of progression and invasion of PC-3 cells. From Western blotting results, the effects of ET-1 regulation and Endostar on PC-3 cells were at least related to some signaling pathways involving PI3K/Akt/Caspase-3, Erk1/2/Bcl-2/Caspase-3 and MMPs (MMP-2 and MMP-9). Furthermore, combined treatment of ET-1RNAi and Endostar was found to be more effective than single treatment. Both ET-1 RNAi and Endostar can inhibit the progression and invasion of PC-3 cells, but combined treatment might be a better therapeutic schedule.
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Pricing and availability is not currently available.