Merck
  • A DNA-scaffolded silver nanocluster/Cu²⁺ ensemble as a turn-on fluorescent probe for histidine.
A DNA-scaffolded silver nanocluster/Cu²⁺ ensemble as a turn-on fluorescent probe for histidine.
The Analyst (2014-05-03)
Ying Zhou, Tianshu Zhou, Min Zhang, Guoyue Shi
ABSTRACT
A new type of rapid, sensitive, and selective fluorescence turn-on assay was developed for detection of histidine using a DNA-scaffolded silver nanocluster/Cu(2+) ensemble (DNA-AgNC/Cu(2+)). Cu(2+) was first bound to nucleic acids of the DNA-AgNCs forming a DNA-AgNC/Cu(2+) ensemble and then liberated into solution via the highly specific interaction between histidine and Cu(2+) in the presence of other amino acids. The fluorescence of DNA-AgNCs was greatly quenched with the addition of Cu(2+), and then the DNA-AgNC/Cu(2+) ensemble exhibited marvelous fluorescent enhancement in the presence of histidine, which showed the possibility for constructing a turn-on chemosensor of histidine. Compared to other methods, this approach promises high sensitivity, simplicity in design, convenient operation, and minimized use of organic solvents. The ultra-high selectivity demonstrated the feasibility of the assay for detecting histidine in sophisticated physical environments. The fitting range of the proposed assay is from 0 to 100 μM, with a detection limit of 1.4 μM (S/N = 3) in artificial urine samples. The protocol was evaluated by analysis of artificial urine samples with good recoveries and showed great potential for practical applications.
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
VenPure® SF, powder
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Alanine, European Pharmacopoeia (EP) Reference Standard
Proline, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
Valine, European Pharmacopoeia (EP) Reference Standard
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Leucine, European Pharmacopoeia (EP) Reference Standard
Isoleucine, European Pharmacopoeia (EP) Reference Standard
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)