Merck
  • Stereoselective skin anti-photoaging properties of ginsenoside Rg3 in UV-B-irradiated keratinocytes.
Stereoselective skin anti-photoaging properties of ginsenoside Rg3 in UV-B-irradiated keratinocytes.
Biological & pharmaceutical bulletin (2014-07-25)
Chang-Jin Lim, Woo-Yong Choi, Hyun-Joo Jung
ABSTRACT
Ginsenosides are major bioactive constituents that are responsible for the diverse pharmacological activities of ginseng. This work aimed to assess the skin anti-photoaging activities of the two stereoisomeric forms of ginsenoside Rg3, 20(S)-Rg3 and 20(R)-Rg3. When the two Rg3 stereoisomers were added to cultured human keratinocyte HaCaT cells prior to irradiation with 70 mJ/cm(2) UV-B, 20(S)-Rg3, but not 20(R)-Rg3, decreased the UV-B-induced intracellular reactive oxygen species (ROS) levels in a concentration-dependent manner, as detected by both fluorometric and confocal microscopic analyses. Likewise, 20(S)-Rg3, but not 20(R)-Rg3, decreased the UV-B-induced ROS levels in human dermal fibroblast cells. Both stereoisomers were unable to modulate the nitric oxide levels in HaCaT cells under UV-B irradiation, and induced no cytotoxicity in cultured keratinocytes and fibroblasts. 20(S)-Rg3 suppressed the UV-B-induced matrix metalloproteinase (MMP)-2 activities in HaCaT cells. Taken together, these results indicate that 20(S)-Rg3 possesses both ROS-scavenging and MMP-2 inhibitory activities, while 20(R)-Rg3 possesses neither activity. These findings imply that ginsenoside Rg3 stereoselectively demonstrates skin anti-photoaging activities.
Sigma-Aldrich
Sodium nitrite, JIS special grade, ≥98.5%
Sigma-Aldrich
Sodium nitrite, SAJ first grade, ≥97.0%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%