Merck
  • Family with sequence similarity 5, member C (FAM5C) increases leukocyte adhesion molecules in vascular endothelial cells: implication in vascular inflammation.
Family with sequence similarity 5, member C (FAM5C) increases leukocyte adhesion molecules in vascular endothelial cells: implication in vascular inflammation.
PloS one (2014-09-25)
Junya Sato, Mitsuo Kinugasa, Seimi Satomi-Kobayashi, Kinta Hatakeyama, Aaron J Knox, Yujiro Asada, Margaret E Wierman, Ken-ichi Hirata, Yoshiyuki Rikitake
ABSTRACT
Identification of the regulators of vascular inflammation is important if we are to understand the molecular mechanisms leading to atherosclerosis and consequent ischemic heart disease, including acute myocardial infarction. Gene polymorphisms in family with sequence similarity 5, member C (FAM5C) are associated with an increased risk of acute myocardial infarction, but little is known about the function of this gene product in blood vessels. Here, we report that the regulation of the expression and function of FAM5C in endothelial cells. We show here that FAM5C is expressed in endothelial cells in vitro and in vivo. Immunofluorescence microscopy showed localization of FAM5C in the Golgi in cultured human endothelial cells. Immunohistochemistry on serial sections of human coronary artery showed that FAM5C-positive endothelium expressed intercellular adhesion molecule-1 (ICAM-1) or vascular cell adhesion molecule-1 (VCAM-1). In cultured human endothelial cells, the overexpression of FAM5C increased the reactive oxygen species (ROS) production, nuclear factor-κB (NF-κB) activity and the expression of ICAM-1, VCAM-1 and E-selectin mRNAs, resulting in enhanced monocyte adhesion. FAM5C was upregulated in response to inflammatory stimuli, such as TNF-α, in an NF-κB- and JNK-dependent manner. Knockdown of FAM5C by small interfering RNA inhibited the increase in the TNF-α-induced production of ROS, NF-κB activity and expression of these leukocyte adhesion molecule mRNAs, resulting in reduced monocyte adhesion. These results suggest that in endothelial cells, when FAM5C is upregulated in response to inflammatory stimuli, it increases the expression of leukocyte adhesion molecules by increasing ROS production and NF-κB activity.
Product Description
SAFC
HEPES
Sigma-Aldrich
MISSION® esiRNA, targeting human BRINP3
SAFC
HEPES
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fam5c
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
2-Thenoyltrifluoroacetone, 99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, JIS special grade, ≥95.0%
Sigma-Aldrich
Calcium chloride, SAJ first grade, ≥95.0%
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, ≥75%, for drying