Merck
  • CADM1 expression and stepwise downregulation of CD7 are closely associated with clonal expansion of HTLV-I-infected cells in adult T-cell leukemia/lymphoma.
CADM1 expression and stepwise downregulation of CD7 are closely associated with clonal expansion of HTLV-I-infected cells in adult T-cell leukemia/lymphoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-04-15)
Seiichiro Kobayashi, Kazumi Nakano, Eri Watanabe, Tomohiro Ishigaki, Nobuhiro Ohno, Koichiro Yuji, Naoki Oyaizu, Satomi Asanuma, Makoto Yamagishi, Tadanori Yamochi, Nobukazu Watanabe, Arinobu Tojo, Toshiki Watanabe, Kaoru Uchimaru
ABSTRACT
Cell adhesion molecule 1 (CADM1), initially identified as a tumor suppressor gene, has recently been reported to be ectopically expressed in primary adult T-cell leukemia-lymphoma (ATL) cells. We incorporated CADM1 into flow-cytometric analysis to reveal oncogenic mechanisms in human T-cell lymphotrophic virus type I (HTLV-I) infection by purifying cells from the intermediate stages of ATL development. We isolated CADM1- and CD7-expressing peripheral blood mononuclear cells of asymptomatic carriers and ATLs using multicolor flow cytometry. Fluorescence-activated cell sorted (FACS) subpopulations were subjected to clonal expansion and gene expression analysis. HTLV-I-infected cells were efficiently enriched in CADM1(+) subpopulations (D, CADM1(pos)CD7(dim) and N, CADM1(pos)CD7(neg)). Clonally expanding cells were detected exclusively in these subpopulations in asymptomatic carriers with high proviral load, suggesting that the appearance of D and N could be a surrogate marker of progression from asymptomatic carrier to early ATL. Further disease progression was accompanied by an increase in N with a reciprocal decrease in D, indicating clonal evolution from D to N. The gene expression profiles of D and N in asymptomatic carriers showed similarities to those of indolent ATLs, suggesting that these subpopulations represent premalignant cells. This is further supported by the molecular hallmarks of ATL, that is, drastic downregulation of miR-31 and upregulation of abnormal Helios transcripts. The CADM1 versus CD7 plot accurately reflects disease progression in HTLV-I infection, and CADM1(+) cells with downregulated CD7 in asymptomatic carriers have common properties with those in indolent ATLs.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)