Merck
  • Bacterial communities associated with aerobic degradation of polybrominated diphenyl ethers from river sediments.
Bacterial communities associated with aerobic degradation of polybrominated diphenyl ethers from river sediments.
Environmental science and pollution research international (2014-10-02)
Chu-Wen Yang, Huang-Wen Huang, Wei-Liang Chao, Bea-Ven Chang
ABSTRACT
Polybrominated diphenyl ethers (PBDEs) are persistent organic pollutants and have therefore drawn much environmental concern. We aimed to compare aerobic degradation of different PBDE congeners under various treatments and reveal the bacterial community associated with PBDE degradation in sediment. Results of this study indicate that degradation rates of BDE-15 were enhanced 45.1 and 81.3 % with the addition of suspended and microencapsulated Pseudomonas sp., respectively. However, the degradation rates of BDE-28, BDE-47, BDE-99, and BDE-100 did not differ among experimental treatments. Degradation rates of PBDE congeners were in the order of BDE-15 > BDE-28 > BDE-47 > BDE-99 > BDE-100. Using a pyrosequencing-based metagenomic approach, we found that addition of various treatments altered the microbial community composition in the sediment. Twenty-four bacterial genera associated with degradation of PBDEs; six are the core bacterial genera common among PBDE degraders. The diverse bacterial composition among different PBDE congener degradation indicates different combinations of bacteria involved in degradation of different PBDE congeners.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)