Merck
  • Nasal fluid release of eotaxin-3 and eotaxin-2 in persistent sinonasal eosinophilic inflammation.
Nasal fluid release of eotaxin-3 and eotaxin-2 in persistent sinonasal eosinophilic inflammation.
International forum of allergy & rhinology (2014-07-06)
Eugenio De Corso, Silvia Baroni, Mariapina Battista, Matteo Romanello, Romina Penitente, Walter Di Nardo, Giulio Cesare Passali, Bruno Sergi, Anna Rita Fetoni, Francesco Bussu, Cecilia Zuppi, Gaetano Paludetti
ABSTRACT
The aim of the present study was to measure eotaxin-3 (CCL26) and eotaxin-2 (CCL24) in nasal lavage fluid of patients with different forms of chronic sinonasal eosinophilic inflammation to evaluate their role in the pathophysiology of nasal hypereosinophilia. The study was an analytic cross-section study, level of evidence 3b. Patients (n = 80) with nasal hypereosinophilia were randomly recruited and grouped in the following categories: persistent allergic rhinitis (AR) (n = 25), nonallergic rhinitis with eosinophilia syndrome (NARES) (n = 30), and chronic rhinosinusitis with polyps (CRSwNP) (n = 25). Non-rhinitic volunteers (n = 20) were recruited as controls. CCL24 and CCL26 concentrations were measured by enzyme-linked immunosorbent assay (ELISA) Quantikine Human Immunoassays (R&D Systems, Minneapolis, MN) in nasal lavage fluids. Differential cell counts were performed by microscopic cytological examination of nasal tissue scraped from the inferior turbinate. Mean CCL26 levels were significantly higher (p < 0.05) in AR and in NARES (132.0 pg/mL and 187.63 pg/mL, respectively) than in the control group (13.5 pg/mL); in patients with CRSwNP, CCL26 values were increased compared to controls even though the difference was not statistically significant (58.9 pg/mL vs 16.5 pg/mL). Mean CCL24 levels measured in AR, NARES, and CRSwNP were significantly increased (p < 0.05) compared to controls (96.7 pg/mL, 135.4 pg/mL, and 107.0 pg/mL, respectively, vs 32.2 pg/mL). Moreover, we observed a significant correlation between CCL24 and CCL26 levels, evaluating them intraindividually by Spearman's rank correlation test. Finally, a significant correlation was found between CCL24 and CCL26 levels and the percentage of eosinophilic infiltration of nasal mucosa. Our data suggest that CCL26 and CCL24 are likely involved in the pathogenesis of chronic nasal hypereosinophilia, with a complex cooperation and different involvement of the various members of eotaxin family. Further studies are necessary to better understand the actual physiopathologic mechanism, possible clinical relevance, and therapeutic implications.
Sigma-Aldrich
Mouse Eotaxin-2 ELISA Kit, for serum, plasma and cell culture supernatant
Pricing and availability is not currently available.
Sigma-Aldrich
Human Eotaxin-2 / CCL24 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Pricing and availability is not currently available.