Merck
  • Effect of anion structures on cholinium ionic liquids pretreatment of rice straw and the subsequent enzymatic hydrolysis.
Effect of anion structures on cholinium ionic liquids pretreatment of rice straw and the subsequent enzymatic hydrolysis.
Biotechnology and bioengineering (2014-07-30)
Xue-Dan Hou, Jie Xu, Ning Li, Min-Hua Zong
ABSTRACT
In this work, 28 cholinium ionic liquids (ILs), most of which are good solvents for dissolving lignin, were used for rice straw pretreatment to improve subsequent enzymatic hydrolysis. The anion exerted a significant effect on the pretreatment effectiveness of the IL as well as the subsequent enzymatic hydrolysis efficiency of rice straw residues. The presence of the basic group(s) in the anion significantly enhanced the IL pretreatment effectiveness, while the carboxyl, hydroxyl and aromatic groups had a negative impact on IL delignification. Except for amino acid-based ILs, the delignification abilities of the ILs are linearly and positively correlated with the pKa values of the conjugate acids of the anions. Of the ILs tested, amino acid-based ILs, especially basic amino acid-based ILs, are the most effective pretreatment solvents. Satisfactory reducing sugar yields (81% for glucose and 26% for xylose) were obtained in the enzymatic hydrolysis of rice straw pretreated by cholinium argininate ([Ch][Arg]) under a pretty mild pretreatment severity (60°C, 6 h). The results presented in this work may be useful for rational design of novel and green ILs for delignification of lignocellulose.
Sigma-Aldrich
Choline hydroxide solution, 46 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Choline hydroxide solution, 45 wt. % in methanol
Pricing and availability is not currently available.