Merck
  • Highly sensitive analysis of flavonoids by zwitterionic microemulsion electrokinetic chromatography coupled with light-emitting diode-induced fluorescence detection.
Highly sensitive analysis of flavonoids by zwitterionic microemulsion electrokinetic chromatography coupled with light-emitting diode-induced fluorescence detection.
Journal of chromatography. A (2014-07-23)
Wan Cao, Shuai-Shuai Hu, Xing-Ying Li, Xiao-Qing Pang, Jun Cao, Li-Hong Ye, Han-Bin Dai, Xiao-Juan Liu, Jian-Hua Da, Chu Chu
ABSTRACT
A rapid zwitterionic microemulsion electrokinetic chromatography (ZI-MEEKC) approach coupled with light-emitting-diode-induced fluorescence (LED-IF, 480nm) detection was proposed for the analysis of flavonoids. In the optimization process, we systematically investigated the separation conditions, including the surfactants, cosurfactants, pH, buffers and fluorescence parameters. It was found that the baseline separation of the seven flavonoids was obtained in less than 5min with a running buffer consisting of 92.9% (v/v) 5mM sodium borate, 0.6% (w/v) ZI surfactant, 0.5% (w/v) ethyl acetate and 6.0% (w/v) 1-butanol. High sensitivity was obtained by the application of LED-IF detection. The limits of detection for seven flavonoids were in the range of 3.30×10(-8) to 2.15×10(-6)molL(-1) without derivatization. Ultimately, the detection method was successfully applied to the analysis of flavonoids in hawthorn plant and food products with satisfactory results.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Supelco
Apigenin, analytical standard
Sigma-Aldrich
3-(N,N-Dimethylpalmitylammonio)propanesulfonate, ≥98% (TLC)
Hyperoside, primary reference standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
3-(N,N-Dimethylmyristylammonio)propanesulfonate, ≥98.0% (TLC)
Sigma-Aldrich
Isovitexin, ≥98.0% (HPLC)
Isoquercitrin, primary reference standard
Supelco
Kaempferide, analytical standard
Supelco
Quercetin 3-glucoside, analytical standard