Merck
  • Culture supernatants of different colon cancer cell lines induce specific phenotype switching and functional alteration of THP-1 cells.
Culture supernatants of different colon cancer cell lines induce specific phenotype switching and functional alteration of THP-1 cells.
Cellular immunology (2014-06-25)
Tsung-Han Wu, Ying-Ying Li, Tai-Ling Wu, John W-C Chang, Wen-Chi Chou, Ling-Ling Hsieh, Jim-Ray Chen, Kun-Yun Yeh
ABSTRACT
We developed an in vitro model to evaluate the effect of products secreted from different colorectal cancer (CRC) cell lines on specific phenotypic switching and functional alterations in THP-1 cells. We co-cultured the human monocytic cell line, THP-1, or phorbol-12-myristate-13-acetate (PMA)-treated THP-1 cells, (THP-1p), with supernatants from either the HT-29 (Dukes' B), HCT-15 (Dukes' C), or Colo205 (Dukes' D) cell lines, and assessed the cells for macrophage differentiation. The surface marker and cytokine profiles suggested that secreted CRC factors differentiated THP-1 cells into a "mixed" M1/M2 phenotype, although HT-29 and Colo205 supernatants induced THP-1p cells into predominantly M1-like macrophages and M2-like macrophages, respectively. Further, all three CRC supernatants enhanced the phagocytic capacity and migration of THP-1 and THP-1p cells, altering their phenotype to a more M2-kind. Therefore, different CRC cell lines induced specific phenotype switching and functional polarization of THP-1 cells.
Sigma-Aldrich
ANTI-CD106 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution