Merck
  • Transferring lithium ions in nanochannels: a PEO/Li⁺ solid polymer electrolyte design.
Transferring lithium ions in nanochannels: a PEO/Li⁺ solid polymer electrolyte design.
Angewandte Chemie (International ed. in English) (2014-03-07)
Ling-Yun Yang, Da-Xiu Wei, Min Xu, Ye-Feng Yao, Qun Chen
ABSTRACT
A new category of crystalline polymer electrolyte prepared by the supramolecular self-assembly of polyethylene oxide (PEO), α-cyclodextrin (α-CD), and LiAsF6 is reported. The polymer electrolyte consists of the nanochannels formed by α-CDs in which the PEO/Li(+) complexes are confined. The nanochannels formed by α-CD provide the pathway for the directional motion of Li(+) ions and at the same time prevent the access of the anions by size exclusion, resulting in good separation of the Li(+) ions and the anions. The conductivity of the reported material is 30 times higher than that of the comparable PEO/Li(+) complex crystal at room temperature. By using state-of-art solid-state NMR spectroscopy, the structure and dynamics of the material were investigated in detail. The dynamics of the Li(+) ions was studied and correlated to the ionic conductivity of the material.
Sigma-Aldrich
Lithium hexafluoroarsenate(V), 98%
Pricing and availability is not currently available.