Merck
  • Molecular basis for zinc transporter 1 action as an endogenous inhibitor of L-type calcium channels.
Molecular basis for zinc transporter 1 action as an endogenous inhibitor of L-type calcium channels.
The Journal of biological chemistry (2009-09-22)
Shiri Levy, Ofer Beharier, Yoram Etzion, Merav Mor, Liat Buzaglo, Lior Shaltiel, Levi A Gheber, Joy Kahn, Anthony J Muslin, Amos Katz, Daniel Gitler, Arie Moran
ABSTRACT
The L-type calcium channel (LTCC) has a variety of physiological roles that are critical for the proper function of many cell types and organs. Recently, a member of the zinc-regulating family of proteins, ZnT-1, was recognized as an endogenous inhibitor of the LTCC, but its mechanism of action has not been elucidated. In the present study, using two-electrode voltage clamp recordings in Xenopus oocytes, we demonstrate that ZnT-1-mediated inhibition of the LTCC critically depends on the presence of the LTCC regulatory beta-subunit. Moreover, the ZnT-1-induced inhibition of the LTCC current is also abolished by excess levels of the beta-subunit. An interaction between ZnT-1 and the beta-subunit, as demonstrated by co-immunoprecipitation and by fluorescence resonance energy transfer, is consistent with this result. Using surface biotinylation and total internal reflection fluorescence microscopy in HEK293 cells, we show a ZnT-1-dependent decrease in the surface expression of the pore-forming alpha(1)-subunit of the LTCC. Similarly, a decrease in the surface expression of the alpha(1)-subunit is observed following up-regulation of the expression of endogenous ZnT-1 in rapidly paced cultured cardiomyocytes. We conclude that ZnT-1-mediated inhibition of the LTCC is mediated through a functional interaction of ZnT-1 with the LTCC beta-subunit and that it involves a decrease in the trafficking of the LTCC alpha(1)-subunit to the surface membrane.