Merck
α-Synuclein modulates neurite outgrowth by interacting with SPTBN1.
Biochemical and biophysical research communications (2012-07-10)
Hak Joo Lee, Kyunghee Lee, Hana Im
ABSTRACT
α-Synuclein is the major component of Lewy bodies and Lewy neurites, the pathological hallmarks of surviving neuronal cells in Parkinson's disease patients. However, the physiological role played by α-synuclein remains unclear. In this study, spectrin beta non-erythrocyte 1 (SPTBN1) interacted with α-synuclein in phage display assays using a normalized human brain cDNA library. A direct interaction between α-synuclein and SPTBN1 was confirmed by GST pull-down and co-immunoprecipitation assays. SPTBN1 and α-synuclein proteins colocalized in N2a neuronal cells. Transfection of SPTBN1 caused human SH-SY5Y dopaminergic neuron cells to inappropriately induce neurites, which extended from cell bodies. Cotransfection with α-synuclein reversed SPTBN1-induced excessive neurite branching in SH-SY5Y cells, and only a single neurite extended from each neuron. These results suggest that α-synuclein modulates neurite outgrowth by interacting with cytoskeletal proteins such as SPTBN1.