- Synthesis and vibrational spectrum of antimony phosphate, SbPO4.
Synthesis and vibrational spectrum of antimony phosphate, SbPO4.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2002-08-08)
Wolfgang Brockner, Lars P Hoyer
PMID12164490
ABSTRACT
SbPO4 was synthesized via a new route by reacting antimony metal with meta-phosphoric acid, (HPO3)n at high temperatures. The Raman and IR spectra of the title compound were recorded and the vibrational modes assigned on the basis of a factor group analysis. The internal vibrations are derived from tetrahedral PO4 units (approaching Sb[PO4]) by the correlation method, although the structure is polymeric and not ionic.
MATERIALS