Merck
  • Ratiometric pH-nanosensors based on rhodamine-doped silica nanoparticles functionalized with a naphthalimide derivative.
Ratiometric pH-nanosensors based on rhodamine-doped silica nanoparticles functionalized with a naphthalimide derivative.
Journal of colloid and interface science (2009-08-15)
Tristan Doussineau, Sabine Trupp, Gerhard J Mohr
ABSTRACT
This paper describes the preparation of two-dye-doped silica nanoparticles for ratiometric pH measurements in the biologically relevant pH-range. While a rhodamine derivative is embedded in a silica core and used as the reference, a pH-sensitive naphthalimide dye is immobilized on the previously amino-functionalized core through two different approaches. Either the naphthalimide's carboxylic group is activated to a succinimidyl-ester to form an amide bond or the system can be built up via solid-phase organic synthesis in only two steps. Both types of nanosensors are characterized in terms of morphology (dynamic light scattering, transmission electron microscopy) and optical properties (steady-state fluorescence spectroscopy). In terms of application, e.g. reproducibility and handling of the synthesis, the first approach gave very good results with respect to size and size distribution and a pK(a) value of 6.55 was found that is comparable to the free indicator dye in solution. The solid-phase organic synthesis method proves the possibility of covalent immobilization of naphthalimides to amino-functionalized surfaces, showing the stability of the polymeric substrate and achieving comparable results for pH sensing.
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Pricing and availability is not currently available.