Merck
Coumarin 1,4-dihydropyridine derivatives.
Bioorganic & medicinal chemistry (1998-07-29)
P Valenti, A Rampa, R Budriesi, A Bisi, A Chiarini
ABSTRACT
A series of 1,4-dihydropyridines bearing a coumarin moiety in 4-position was synthesized. The compounds were evaluated for inotropic, chronotropic and calcium antagonist activities. The replacement of the o-nitrophenyl moiety of nifedipine with a coumarin or phenylcoumarin system is accompanied by a decrease of the activity on myocardial and vascular parameters, but the synthesized compounds showed selective inhibiting effects on cardiac contractility and frequency.
Sigma-Aldrich
2-Hydroxy-3-methylbenzaldehyde, 98%
Pricing and availability is not currently available.