Merck
  • sn-glycerol-1-phosphate dehydrogenase in Methanobacterium thermoautotrophicum: key enzyme in biosynthesis of the enantiomeric glycerophosphate backbone of ether phospholipids of archaebacteria.
sn-glycerol-1-phosphate dehydrogenase in Methanobacterium thermoautotrophicum: key enzyme in biosynthesis of the enantiomeric glycerophosphate backbone of ether phospholipids of archaebacteria.
Journal of biochemistry (1995-05-01)
M Nishihara, Y Koga
ABSTRACT
One of the most characteristic features of archaebacterial ether phospholipids is the enantiomeric configuration of their glycerophosphate backbone (sn-glycerol-1-phosphate), that is the mirror image of the structure of the eubacterial or eukaryotic counterpart. The enzyme that forms glycerophosphate of this configuration was found for the first time in a cell-free extract of the methanogen, Methanobacterium thermoautotrophicum, and was identified as sn-glycerol-1-phosphate:NAD+ oxidoreductase (sn-glycerol-1-phosphate dehydrogenase) after partial purification. Because sn-glycerol-1-phosphate has been found to be a precursor of ether lipids of this organism, sn-glycerol-1-phosphate dehydrogenase is a key enzyme in the biosynthesis of the enantiomeric ether lipids of methanogens.
Sigma-Aldrich
sn-Glycerol 1-phosphate lithium salt, ≥95.0% (TLC)