Merck
  • Synthesis, spectral and antimicrobial studies of bis(cyclopentadienyl)titanium(IV) derivatives with Schiff bases derived from 2-amino-5-phenyl-1,3,4-thiadiazole.
Synthesis, spectral and antimicrobial studies of bis(cyclopentadienyl)titanium(IV) derivatives with Schiff bases derived from 2-amino-5-phenyl-1,3,4-thiadiazole.
Bioinorganic chemistry and applications (2008-03-28)
A K Srivastava, O P Pandey, S K Sengupta
ABSTRACT
The reactions of bis(cyclopentadienyl)titanium(IV) dichloride with Schiff bases derived by condensing 2- amino-5-phenyl-1,3,4-thiadiazole with benzaldehyde (SPT), 4-nitrobenzaldehyde (SNT), 4-methoxybenzaldehyde (SMT), 2-hydroxybenzaldehyde (SSTH) or 2-hydroxyacetophenone (SATH) have been studied in refluxing tetrahydrofuran and complexes of types [Cp(2)TiCl(SB)]Cl (SB= SPT, SNT or SMT) and [Cp(2)Ti(SB')]Cl (SB'H= SSTH or SATH) have been isolated. Tentative structural conclusions are drawn for these reaction products based upon elemental analyses, electrical conductance, magnetic moment and spectral (UV-vis, IR and (1)H NMR) data. Studies were conducted to assess the growth-inhibiting potential of the complexes synthesized, and the ligands, against various fungal and bacterial strains.
Sigma-Aldrich
2-Amino-5-phenyl-1,3,4-thiadiazole, 96%
Pricing and availability is not currently available.