Merck
  • Determination of neurotoxin 3-N-oxalyl-2,3-diaminopropionic acid and non-protein amino acids in Lathyrus sativus by precolumn derivatization with 1-fluoro-2,4-dinitrobenzene.
Determination of neurotoxin 3-N-oxalyl-2,3-diaminopropionic acid and non-protein amino acids in Lathyrus sativus by precolumn derivatization with 1-fluoro-2,4-dinitrobenzene.
Journal of chromatography. A (2000-07-26)
F Wang, X Chen, Q Chen, X Qin, Z Li
ABSTRACT
A rapid and simple method is presented for determining neuro-excitatory nonprotein amino acid 3-N-oxalyl-2,3-diaminopropionic acid (beta-ODAP) and non-protein amino acids in Lathyrus sativus. Seed and foliage extracts of Lathyrus sativus were treated with 1-fluoro-2,4-dinitrobenzene (FDNB) and a reversed-phase high-performance liquid chromatography method (RP HPLC) for the separation of the derivatives in the pmol range is reported. The RP HPLC method and a colorimetric method were compared for measuring ODAP.
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, purum p.a., ≥98.0% (GC)
Pricing and availability is not currently available.