Merck
  • Tosvinyl and besvinyl as protecting groups of imides, azinones, nucleosides, sultams, and lactams. Catalytic conjugate additions to tosylacetylene.
Tosvinyl and besvinyl as protecting groups of imides, azinones, nucleosides, sultams, and lactams. Catalytic conjugate additions to tosylacetylene.
The Journal of organic chemistry (2014-08-28)
Elena Petit, Lluís Bosch, Joan Font, Laura Mola, Anna M Costa, Jaume Vilarrasa
ABSTRACT
The use of the 2-(4-methylphenylsulfonyl)ethenyl (tosvinyl, Tsv) group for the protection of the NH group of a series of imides, azinones (including AZT), inosines, and cyclic sulfonamides has been examined. The Tsv-protected derivatives are obtained in excellent yields by conjugate addition to tosylacetylene (ethynyl p-tolyl sulfone). The stereochemistry of the double bond can be controlled at will: with only 1 mol % of Et3N or with catalytic amounts of NaH, the Z stereoisomers are generated almost exclusively, while the E isomers are obtained using a stoichiometric amount of DMAP. Analogous phenylsulfonylvinyl-protected groups (with the besvinyl or Bsv group instead of Tsv) are obtained stereospecifically by reaction with (Z)- or (E)-bis(phenylsulfonyl)ethene. For lactams and oxazolidinones, this last method is much better. The Tsv and Bsv groups are stable in the presence of non-nucleophilic bases and to acids. They can be removed highly effectively via a conjugate addition-elimination mechanism using pyrrolidine or sodium dodecanethiolate as nucleophiles.
Sigma-Aldrich
Ethynyl p-tolyl sulfone, 98%
Pricing and availability is not currently available.