- Novel chiral "calixsalen" macrocycle and chiral robson-type macrocyclic complexes.
Novel chiral "calixsalen" macrocycle and chiral robson-type macrocyclic complexes.
Inorganic chemistry (2005-01-18)
Jian Gao, Joseph H Reibenspies, Ralph A Zingaro, F Ross Woolley, Arthur E Martell, Abraham Clearfield
PMID15651868
ABSTRACT
A family of novel chiral "calixsalen" Schiff base macrocycles R,R-H(3)L4, R,R-H(3)L5, containing three chiral diamino moieties were synthesized by an efficient self-assembly and characterized by (1)H and (13)C NMR, mass spectrometry, and X-ray diffraction. The systematic synthesis, structure, and coordination properties of the [2 + 2] and [3 + 3] Robson-type Schiff base macrocyclic mono-, di-, tri-, and tetranuclear metal complexes were explored.