Merck
  • Investigation in a murine model of possible mechanisms of enhanced local reactions to post-primary diphtheria-tetanus toxoid boosters in recipients of acellular pertussis-diphtheria-tetanus vaccine.
Investigation in a murine model of possible mechanisms of enhanced local reactions to post-primary diphtheria-tetanus toxoid boosters in recipients of acellular pertussis-diphtheria-tetanus vaccine.
Human vaccines & immunotherapeutics (2014-11-27)
Masaki Ochiai, Yoshinobu Horiuchi, Chun-Ting Yuen, Catpagavalli Asokanathan, Akihiko Yamamoto, Kenji Okada, Michiyo Kataoka, Kevin Markey, Michael Corbel, Dorothy Xing
ABSTRACT
In recipients primed with acellular pertussis diphtheria-tetanus combined vaccine (DTaP) an increased incidence of severe local reactions with extensive redness/swelling has been reported for each subsequent dose of diphtheria-tetanus based combination vaccine given as a booster. This has been attributed to residual active pertussis toxin (PT) in the primary vaccine. In this study, we investigated the possible contribution of the A-subunit enzymatic activity and the B-oligomer carbohydrate binding activity of residual PT in DTaP to local reactions in a murine model using Japanese DTaP batches produced before and after the introduction of a test for reversion of pertussis toxoid to toxin. Residual PT activity was correlated with the B-oligomer carbohydrate binding activity. The in vivo mouse footpad swelling model assay indicated that the B-oligomer carbohydrate binding activity and possibly other factors were associated with intensified sensitization to local reaction following diphtheria toxoid booster.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody
Pricing and availability is not currently available.