Merck
  • The ratio of acetate-to-glucose oxidation in astrocytes from a single 13C NMR spectrum of cerebral cortex.
The ratio of acetate-to-glucose oxidation in astrocytes from a single 13C NMR spectrum of cerebral cortex.
Journal of neurochemistry (2014-09-19)
Isaac Marin-Valencia, M Ali Hooshyar, Kumar Pichumani, A Dean Sherry, Craig R Malloy
ABSTRACT
The (13) C-labeling patterns in glutamate and glutamine from brain tissue are quite different after infusion of a mixture of (13) C-enriched glucose and acetate. Two processes contribute to this observation, oxidation of acetate by astrocytes but not neurons, and preferential incorporation of α-ketoglutarate into glutamate in neurons, and incorporation of α-ketoglutarate into glutamine in astrocytes. The acetate:glucose ratio, introduced previously for analysis of a single (13) C NMR spectrum, provides a useful index of acetate and glucose oxidation in the brain tissue. However, quantitation of relative substrate oxidation at the cell compartment level has not been reported. A simple mathematical method is presented to quantify the ratio of acetate-to-glucose oxidation in astrocytes, based on the standard assumption that neurons do not oxidize acetate. Mice were infused with [1,2-(13) C]acetate and [1,6-(13) C]glucose, and proton decoupled (13) C NMR spectra of cortex extracts were acquired. A fit of those spectra to the model indicated that (13) C-labeled acetate and glucose contributed approximately equally to acetyl-CoA (0.96) in astrocytes. As this method relies on a single (13) C NMR spectrum, it can be readily applied to multiple physiologic and pathologic conditions. Differences in (13) C labeling of brain glutamate and glutamine have been attributed to metabolic compartmentation. The acetate:glucose ratio, introduced for description of a (13) C NMR (nuclear magnetic resonance) spectrum, is an index of glucose and acetate oxidation in brain tissue. A simple mathematical method is presented to quantify the ratio of acetate-to-glucose oxidation in astrocytes from a single NMR spectrum. As kinetic analysis is not required, the method is readily applicable to analysis of tissue extracts. α-KG = alpha-ketoglutarate; CAC = citric acid cycle; GLN = glutamine; GLU = glutamate.
Sigma-Aldrich
α-Ketoglutaric acid potassium salt, ≥98% (enzymatic)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, BioUltra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, ≥98% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
SKU
Pack Size
Availability
Price
Quantity
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid hydrochloride
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Supelco
α-Ketoglutaric acid disodium salt hydrate, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Ketoglutaric acid disodium salt dihydrate, ≥98.0% (dried material, NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SKU
Pack Size
Availability
Price
Quantity
SAFC
L-Glutamine
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland