Merck
  • One-pot chemoenzymatic multicomponent synthesis of thiazole derivatives.
One-pot chemoenzymatic multicomponent synthesis of thiazole derivatives.
Molecules (Basel, Switzerland) (2013-11-02)
Hui Zheng, Yi-Jia Mei, Kui Du, Xian-Ting Cao, Peng-Fei Zhang
ABSTRACT
A novel chemoenzymatic one-pot multicomponent synthesis of thiazole derivatives was developed. A series of thiazole derivatives were synthesized with high yields up to 94% under mild enzyme-catalyzed conditions. The blank and control experiments reveal that trypsin from porcine pancreas (PPT) displayed great catalytic activity to promote this reaction and showed a wide tolerance range towards different substrate amines. This trypsin-catalyzed multicomponent conversion method provides a novel strategy to synthesize thiazole derivatives and expands the application of enzymes in organic synthesis.
Sigma-Aldrich
Trypsin from porcine pancreas, BioReagent, Proteomics Grade, Dimethylated