Merck
  • Outcomes and predictive factors of prostate cancer patients with extremely high prostate-specific antigen level.
Outcomes and predictive factors of prostate cancer patients with extremely high prostate-specific antigen level.
Journal of cancer research and clinical oncology (2014-04-22)
Kouji Izumi, Wen-Jye Lin, Hiroshi Miyamoto, Chiung-Kuei Huang, Aerken Maolake, Yasuhide Kitagawa, Yoshifumi Kadono, Hiroyuki Konaka, Atsushi Mizokami, Mikio Namiki
ABSTRACT
Prostate-specific antigen (PSA) is a useful biomarker of prostate cancer (PCa). High-risk localized PCa is defined using T stage, Gleason score (GS), and PSA. However, PSA level defining high-risk PCa is at most 20 ng/mL. In PCa patients with high PSA, it is unclear whether PSA itself can be a prognostic factor. Of 642 patients who were diagnosed as PCa, 90 patients with PSA > 100 ng/mL were retrospectively analyzed. Patients were divided into three groups according to PSA level: very high (>1,000 ng/mL), moderately high (200-1,000 ng/mL), and slightly high (100-200 ng/mL). There were no significant differences in overall survival or PCa-specific survival (PCaSS) among the three groups. Regardless of PSA level, high M stage and GS significantly reduced PCaSS. When the risk classification was made using M stage and GS (high risk = M1 and GS ≥ 9, low risk = M0 and GS < 9, and intermediate risk = others), PCaSS was significantly different among high-, intermediate-, and low-risk groups with 5-year survival rates of 58.2, 80.6, and 100 %, respectively. Although there were no differences in treatment performed during the castration-resistant stage, patients undergoing alternative anti-androgen and zoledronic acid treatment had better PCaSS after being castration-resistant. As PSA could not be a prognostic factor in PCa patients with high PSA > 100 ng/mL, the novel risk classification using M stage and GS may help clinicians to predict PCaSS and to plan follow-up schedules after diagnosis.