MilliporeSigma
  • Microwave-assisted one-pot synthesis of 1-indanones from arenes and alpha,beta-unsaturated acyl chlorides.
Microwave-assisted one-pot synthesis of 1-indanones from arenes and alpha,beta-unsaturated acyl chlorides.
The Journal of organic chemistry (2006-05-20)
Wei Yin, Yuan Ma, Jiaxi Xu, Yufen Zhao
ABSTRACT
A series of 1-indanones were synthesized in good yields via tandem Friedel-Crafts acylation and Nazarov cyclization of arenes and alpha,beta-unsaturated acyl chlorides in the presence of aluminum chloride under microwave irradiation.