Merck
  • Permethylated beta-cyclodextrin in liquid poly(oxyethylene) as a stationary phase for capillary gas chromatography.
Permethylated beta-cyclodextrin in liquid poly(oxyethylene) as a stationary phase for capillary gas chromatography.
Journal of chromatography. A (2009-09-01)
Jaiver Osorio Grisales, Pablo J Lebed, Sonia Keunchkarian, Francisco R González, Cecilia B Castells
ABSTRACT
This paper reports the study of poly(oxyethylene) as a solvent for heptakis(2,3,6-tri-O-methyl)-beta-cyclodextrin (PM-beta-CD) and the potential of these mixtures for constructing enantioselective gas chromatography columns. Enantioseparations of volatile racemic mixtures using capillaries coated with 10-50% PM-beta-CD diluted in Carbowax 20M were evaluated. The influence of the polymer type on retention, separation and reproducibility over a period of time is critically discussed. The poly(oxyethylene)-based columns were also compared to columns prepared by dilution of PM-beta-CD in two polysiloxanes, SE-30 and SE-54, at several temperatures. Finally, we evaluated a new stationary phase prepared by crosslinking the chiral selector to the poly(oxyethylene) matrix.
Product Description
Supelco
GC Stationary Phase, phase Carbowax 20M, bottle of 50 g