Merck
  • Novel naphthyridines are histamine H3 antagonists and serotonin reuptake transporter inhibitors.
Novel naphthyridines are histamine H3 antagonists and serotonin reuptake transporter inhibitors.
Bioorganic & medicinal chemistry letters (2007-02-20)
Michael A Letavic, John M Keith, Kiev S Ly, Ann J Barbier, Jamin D Boggs, Sandy J Wilson, Brian Lord, Timothy W Lovenberg, Nicholas I Carruthers
ABSTRACT
A series of novel tetrahydronaphthyridine-based histamine H(3) ligands that have serotonin reuptake transporter inhibitor activity is described. The 1,2,3,4-tetrahydro-2,6-naphthyridine scaffold is assembled via the addition of a nitrostyrene to a metalated pyridine followed by reduction and cyclization to form the naphthyridine. In vitro biological data for these novel compounds are discussed.