Merck
  • Activation of natural killer-like YT-INDY cells by oligodeoxynucleotides and binding by homologous pattern recognition proteins.
Activation of natural killer-like YT-INDY cells by oligodeoxynucleotides and binding by homologous pattern recognition proteins.
Scandinavian journal of immunology (2005-10-29)
H Kaur, L Jaso-Friedmann, J H Leary, K Praveen, Z Brahmi, D L Evans
ABSTRACT
The present study was designed to examine the binding and signalling effects of single base and CpG dinucleotide phosphodiester (Po) oligodeoxynucleotides (ODN) on the human natural killer (NK)-like cell line (YT-INDY). Single base Po ODN composed of 20-mers of guanosine (dG20), adenosine (dA20), cytosine (dC20) or thymidine (dT20) as well as 'conventional' Po CpG ODN were examined for their ability to bind and activate YT-INDY cells. Binding by dG20 and CpG ODN to YT-INDY cells was saturable and specific. dG20 binding was competitively inhibited by homologous dG20 and heterologous CpG ODN but not by dC20 and dA20. Two different YT-INDY membrane proteins (18 and 29 kDa) were identified by ligand (Southwestern) blotting with biotinylated dG20 and CpG. The specificity of the ODN-binding protein(s) was further confirmed by ODN depletion experiments using a teleost recombinant protein orthologue [nonspecific cytotoxic cells (NCC) cationic antimicrobial protein-1 (ncamp-1)] known to bind CpG and dG20. Cell proliferation and activation studies showed that dG20 and CpG treatment of YT-INDY cells induced cellular DNA synthesis (i.e. G1 to S-phase conversion). This signalling function was accompanied in dG20-treated cells by proliferation 10 h posttreatment. Both dG20 and CpG ODN binding induced a calcium flux in YT-INDY cells within seconds of treatment. These experiments demonstrated that Po single base dG20 and CpG ODN bind to a (potential) new class of cell-surface proteins that mediate the activation of YT-INDY cells.
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Pricing and availability is not currently available.