Merck
  • Development of a simultaneous liquid chromatography/tandem mass spectrometric method for the determination of type B trichothecenes, their derivatives, and precursors in wheat.
Development of a simultaneous liquid chromatography/tandem mass spectrometric method for the determination of type B trichothecenes, their derivatives, and precursors in wheat.
Rapid communications in mass spectrometry : RCM (2011-11-19)
Tomoyuki Kadota, Makoto Kimura, Satoshi Hirano, Osamu Tajima, Takashi Nakajima, Yoichi Kamata, Yoshiko Sugita-Konishi
ABSTRACT
A method coupling liquid chromatography with tandem mass spectrometry (LC/MS/MS) was developed for the simultaneous quantitative determination of trichothecenes, nivalenol, deoxynivalenol, deoxynivalenol-3-glucoside, fusarenon-X, 3-acetyldeoxynivalenol, 15-acetyldeoxynivalenol, isotrichodermin, calonectrin, 3-deacetylcalonectrin, 15-deacetylcalonectrin, 3,15-diacetylnivalenol, 4,15-diacetylnivalenol, 3,15-diacetyldeoxynivalenol, and 3,4,15-triacetylnivalenol. The analytical parameters of trichothecenes and their derivatives were optimized to enable their highly sensitive detection. Evaluation of clean-up procedures using Multisep #226 and #227 indicated that Multisep #227 was more suitable for their simultaneous detection in wheat. In performance validation studies using the LC/MS/MS method with Multisep #227 cleanup, good recoveries ranging from 84% to 115% with relative standard deviations from 0.4% to 7.2% were measured. The limits of detection and quantification ranged from 0.03 to 1.4 ng·g(-1) and from 0.1 to 4.7 ng·g(-1) , respectively. The effect of matrices using matrix-matched calibration was estimated to range from 80% to 117% after Multisep #227 cleanup. Multisep #227 clean-up procedure with matrix-free standard calibration achieved accurate quantification without having a considerable effect on matrix compounds. Using the developed method, several trichothecene derivatives and precursors were detected in fungally inoculated wheat samples. The developed LC/MS/MS method is a practical technique that can be used for the quantification of trichothecenes in wheat. This study is the first report of an analytical method used for the simultaneous quantification of major trichothecenes, their derivatives and precursors.
Product Description
Supelco
B-Trichothecene mix, (DON, NIV, 3-AcDON, 15-AcDON), ~100 μg/mL in acetonitrile (each of DON, NIV, 3-AcDON and 15-AcDON), analytical standard
Pricing and availability is not currently available.