Merck
  • Synthesis, reactions and biological evaluation of some new naphtho[2,1-b]furan derivatives bearing a pyrazole nucleus.
Synthesis, reactions and biological evaluation of some new naphtho[2,1-b]furan derivatives bearing a pyrazole nucleus.
Molecules (Basel, Switzerland) (2011-01-27)
Ashraf H F Abd El-Wahab, Zarrag Isa A Al-Fifi, Ahmed H Bedair, Fawzy M Ali, Ahmed H A Halawa, Ahemed M El-Agrody
ABSTRACT
Vilsmeier formylation of 2-(1-phenylhydrazonoethyl)naphtho[2,1-b]furan (2) gave 3-naphtho[2,1-b]furan-2-yl-1-phenyl-1H-pyrazole-4-carbaldehyde (3), which was reacted with C- and N-nucleophiles to afford naphthofuranpyrazol derivatives 4-8. Treatment of 2-[(3-(naphtho[2,1-b]furan-2-yl)-1-phenyl-1H-pyrazol-4-yl)methylene]-malononitrile (4a) with reactants having active hydrogen and Et₃N gave the corresponding pyrazoline, pyran and chromene addition product derivatives 10, 12 and 13, consisting of three different connected heterocyclic moieties. Reaction of 1-((3-(naphtho[2,1-b]furan-2-yl)-1-phenyl-1H-pyrazol-4-yl) methylene)-2-phenylhydrazone (6b) with AcONa and ethyl bromoacetate or chloroacetone afforded the thiazolidinone and methylthiazole derivatives 14 and 15, respectively. In addition, intramolecular cyclization of 6d with Ac₂O afford the corresponding 1,3,4-thiadiazol-2-yl acetamide derivative 16. The structures of the synthesized compounds were confirmed by IR, ¹H-NMR/¹³C-NMR and mass spectral studies. Compound 14 showed promising effects against the tested Gram positive and negative bacteria and fungi.
Sigma-Aldrich
Chloroacetone, Wacker Chemie AG, ≥96.0% (GC)