Merck
  • [The role of glipizide in the treatment of non-insulin dependent diabetes].
[The role of glipizide in the treatment of non-insulin dependent diabetes].
Przeglad lekarski (1996-01-01)
M Małecki
ABSTRACT
The structure and effect of glipizide, the second generation derivates of sulfonylurea were described. The special role in the treatment of polymetabolic syndrome of this drug was stressed.
Sigma-Aldrich
Glipizide, solid
Pricing and availability is not currently available.
Glipizide, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.