- [Allergic contact dermatitis caused by etofenamate].
[Allergic contact dermatitis caused by etofenamate].
Zeitschrift fur Hautkrankheiten (1988-06-15)
A Degenhardt, C Zick, B M Hausen
PMID2970161
ABSTRACT
We report on 3 cases of contact dermatitis following topical application of etofenamat. Each patient developed positive reactions in patch tests with etofenamat in concentrations of both 10% and 1%. After testing, one patient showed secondary inflammatory reactions at the original site of application. Considering the frequent administration of etofenamat, contact sensitization seems to be relatively rare.
Product Description
Etofenamate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Etofenamate for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.