Merck
  • Outcomes for efavirenz versus nevirapine-containing regimens for treatment of HIV-1 infection: a systematic review and meta-analysis.
Outcomes for efavirenz versus nevirapine-containing regimens for treatment of HIV-1 infection: a systematic review and meta-analysis.
PloS one (2013-07-31)
Prinitha Pillay, Nathan Ford, Zara Shubber, Rashida A Ferrand
ABSTRACT
There is conflicting evidence and practice regarding the use of the non-nucleoside reverse transcriptase inhibitors (NNRTI) efavirenz (EFV) and nevirapine (NVP) in first-line antiretroviral therapy (ART). We systematically reviewed virological outcomes in HIV-1 infected, treatment-naive patients on regimens containing EFV versus NVP from randomised trials and observational cohort studies. Data sources include PubMed, Embase, the Cochrane Central Register of Controlled Trials and conference proceedings of the International AIDS Society, Conference on Retroviruses and Opportunistic Infections, between 1996 to May 2013. Relative risks (RR) and 95% confidence intervals were synthesized using random-effects meta-analysis. Heterogeneity was assessed using the I(2) statistic, and subgroup analyses performed to assess the potential influence of study design, duration of follow up, location, and tuberculosis treatment. Sensitivity analyses explored the potential influence of different dosages of NVP and different viral load thresholds. Of 5011 citations retrieved, 38 reports of studies comprising 114 391 patients were included for review. EFV was significantly less likely than NVP to lead to virologic failure in both trials (RR 0.85 [0.73-0.99] I(2) = 0%) and observational studies (RR 0.65 [0.59-0.71] I(2) = 54%). EFV was more likely to achieve virologic success than NVP, though marginally significant, in both randomised controlled trials (RR 1.04 [1.00-1.08] I(2) = 0%) and observational studies (RR 1.06 [1.00-1.12] I(2) = 68%). EFV-based first line ART is significantly less likely to lead to virologic failure compared to NVP-based ART. This finding supports the use of EFV as the preferred NNRTI in first-line treatment regimen for HIV treatment, particularly in resource limited settings.
Sigma-Aldrich
Nevirapine
Nevirapine (anhydrous), European Pharmacopoeia (EP) Reference Standard
Nevirapine for peak identification, European Pharmacopoeia (EP) Reference Standard