Merck
Oral contraceptives containing drospirenone for premenstrual syndrome.
The Cochrane database of systematic reviews (2012-02-18)
Laureen M Lopez, Adrian A Kaptein, Frans M Helmerhorst
ABSTRACT
Premenstrual syndrome (PMS) is a common problem. Premenstrual dysphoric disorder (PMDD) is a severe form of premenstrual syndrome. Combined oral contraceptives, which provide both progestin and estrogen, have been examined for their ability to relieve premenstrual symptoms. An oral contraceptive containing drospirenone and a low estrogen dose has been approved for treating PMDD in women who choose oral contraceptives for contraception. To review all randomized controlled trials comparing a combined oral contraceptive containing drospirenone to a placebo or another combined oral contraceptive for effect on premenstrual symptoms. We searched for studies of drospirenone and premenstrual syndrome in the following databases: Cochrane Central Register of Controlled Trials (CENTRAL) (The Cochrane Library), MEDLINE, and POPLINE (20 Dec 2011); EMBASE, LILACS, PsycINFO, ClinicalTrials.gov, and the International Clinical Trials Registry Platform (ICTRP) of the World Health Organization (02 Mar 2011). We also examined references lists of relevant articles and wrote to known investigators to find other trials. We included randomized controlled trials in any language that compared a combined oral contraceptive (COC) containing drospirenone with a placebo or with another COC for effect on premenstrual symptoms. The primary outcome included affective and physical premenstrual symptoms that were prospectively recorded. Adverse events related to combined oral contraceptive use were examined. Two review authors independently extracted data and assessed study quality. For continuous variables, the mean difference (MD) was computed with 95% confidence interval (CI). For dichotomous outcomes, the Peto odds ratio (OR) with 95% CI was calculated. We included five trials with a total of 1920 women. Two placebo-controlled trials of women with PMDD showed less severe premenstrual symptoms after three months with drospirenone 3 mg plus ethinyl estradiol 20 μg than with placebo (MD -7.92; 95% CI -11.16 to -4.67). The drospirenone group had greater mean decreases in impairment of productivity (MD -0.31; 95% CI -0.55 to -0.08), social activities (MD -0.29; 95% CI -0.54 to -0.04), and relationships (MD -0.30; 95% CI -0.54 to -0.06). Side effects more common with the use of the drospirenone COC contraceptive were nausea, intermenstrual bleeding, and breast pain. The respective odds ratios were 3.15 (95% CI 1.90 to 5.22), 4.92 (95% CI 3.03 to 7.96), and 2.67 (95% CI 1.50 to 4.78). Total adverse events related to the study drug were more likely for the drospirenone COC group (OR 2.36; 95% CI 1.62 to 3.44). Three trials studied the effect of drospirenone 3 mg plus ethinyl estradiol 30 μg on less severe symptoms. A placebo-controlled six-month trial had insufficient data for primary outcome analysis. Another six-month study used levonorgestrel 150 µg plus ethinyl estradiol 30 µg for the comparison group but did not provide enough data on premenstrual symptoms. In a two-year trial, the drospirenone COC group had similar premenstrual symptoms to the comparison group given desogestrel 150 µg plus ethinyl estradiol 30 µg (OR 0.87; 95% CI 0.63 to 1.22). The groups were also similar for adverse events related to treatment (OR 1.02; 95% CI 0.78 to 1.33). Drospirenone 3 mg plus ethinyl estradiol 20 μg may help treat premenstrual symptoms in women with severe symptoms, that is, premenstrual dysphoric disorder. The placebo also had a large effect. We do not know whether the combined oral contraceptive works after three cycles, helps women with less severe symptoms, or is better than other oral contraceptives. Larger and longer trials of higher quality are needed to address these issues. Trials should follow CONSORT guidelines.
Product Description
Drospirenone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Drospirenone, ≥98% (HPLC)
Pricing and availability is not currently available.