Merck
  • Pulp response to resin-modified glass ionomer and calcium hydroxide cements in deep cavities: A quantitative systematic review.
Pulp response to resin-modified glass ionomer and calcium hydroxide cements in deep cavities: A quantitative systematic review.
Dental materials : official publication of the Academy of Dental Materials (2010-05-11)
Steffen Mickenautsch, Veerasamy Yengopal, Avijit Banerjee
ABSTRACT
To quantitatively determine whether the pulp response to resin-modified glass ionomer cements placed in deep cavities differs from that generated by calcium hydroxide cement. Five databases were searched for articles up to 31 May 2009. titles/abstracts relevant to topic; published in English; two-arm longitudinal in vivo trial; containing computable dichotomous datasets for test and control group. not all entered subjects accounted for at the end of the trial; subjects of both groups not followed up in the same way; trial on animal tissue. One randomized and five non-randomized control trials, reporting on 1 and 17 datasets, respectively, were accepted. From non-randomized trials, the Relative Risk with 95% Confidence Interval of 13 datasets showed no statistically significant differences (p>0.05) and 4 showed a statistically significant difference between both materials. Meta-analysis of datasets from these trials found no difference between the inflammatory cell response after 30 days (0.87; 95%CI 0.59-1.26; p=0.46); 38% less inflammatory cell response with calcium hydroxide after 60 days (0.62; 95%CI 0.50-0.76; p<0.00001); higher number of intact odontoblasts beneath restored cavities after 381 days (0.56; 95%CI 0.38-0.82; p=0.0008). The results from the randomized control trial (1.40; 95%CI 0.92-2.14; p=0.11) indicated no difference in clinically identifiable pulp symptoms after two years. All trials showed limited internal validity due to selection bias. No conclusive statement about the superiority of either type of material can yet be made. Further high-quality randomized control trials are needed.
Sigma-Aldrich
Calcium hydroxide, CP
Sigma-Aldrich
Calcium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Calcium hydroxide, SAJ special grade, ≥96.0%
Sigma-Aldrich
Calcium hydroxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium hydroxide, ACS reagent, ≥95.0%