Merck
Fragrance material review on cis-nerolidol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review cis-nerolidol when used as a fragrance ingredient is presented.
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans