Merck
  • Analysis of heat-induced contaminants (acrylamide, chloropropanols and furan) in carbohydrate-rich food.
Analysis of heat-induced contaminants (acrylamide, chloropropanols and furan) in carbohydrate-rich food.
Analytical and bioanalytical chemistry (2007-08-04)
Thomas Wenzl, Dirk W Lachenmeier, Vural Gökmen
ABSTRACT
Heat-induced food contaminants have attracted attention of both the scientific community and the public in recent years. The presence of substances considered possibly or probably carcinogenic to humans has triggered an extensive debate on the healthiness of even staple foods. In that respect, acrylamide, furan and chloropropanols are the main substances of concern. Their widespread occurrence in processed food, which concomitantly causes considerable exposure to humans, led either to the setting of maximum limits (for some chloropropanols) or at least the initiation of monitoring programmes in order to put risk assessment on a solid data basis. Acrylamide, furan and chloropropanols are small molecules with physicochemical properties that make their analysis challenging. Their amount in food ranges typically from below the limit of detection to hundreds of micrograms per kilo or even milligrams per kilo. However, a number of recently published scientific reports deal with the analysis of these substances in different kinds of food. The aim of this publication is to give an overview of analytical approaches for the determination of acrylamide, furan and chloropropanols in foodstuffs.
Sigma-Aldrich
Furan, ≥99%