Merck
  • Targeting the methyl erythritol phosphate (MEP) pathway for novel antimalarial, antibacterial and herbicidal drug discovery: inhibition of 1-deoxy-D-xylulose-5-phosphate reductoisomerase (DXR) enzyme.
Targeting the methyl erythritol phosphate (MEP) pathway for novel antimalarial, antibacterial and herbicidal drug discovery: inhibition of 1-deoxy-D-xylulose-5-phosphate reductoisomerase (DXR) enzyme.
Current pharmaceutical design (2007-04-14)
Nidhi Singh, Gweneal Chevé, Mitchell A Avery, Christopher R McCurdy
ABSTRACT
The 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway for isoprenoid biosynthesis has come under increased scrutiny as a target for novel antimalarial, antibacterial and herbicidal agents. 1-Deoxy-D-xylulose 5-phosphate reductoisomerase (DXR) is a key enzyme of the pathway that catalyzes the rearrangement and nicotinamide adenine dinucleotide phosphate (NADPH)-dependent reduction of 1-deoxy-D-xylulose 5-phosphate (DXP) to MEP. The unique properties of DXR make it a remarkable and rational target for drug design. First, it is a vital enzyme for synthesis of isoprenoids in algae, plants, several eubacteria including the pathogenic bacteria like Bacillus anthracis, Helicobacter pylori, Yersinia pestis, Mycobacterium tuberculosis and the malarial parasite, Plasmodium falciparum. Second, there are no functional equivalents to DXR in humans, making it an attractive target for therapeutic intervention. Third, DXR appears to be a valid target and the results from fosmidomycin (1), the only available DXR inhibitor under clinical trials, suggests synergistic effects with the lincosamide antibiotics, lincomycin and clindamycin. Despite drug design efforts in this area, no successful drug specifically designed to inhibit DXR has emerged yet. This review summarizes the recent and promising developments with respect to the current knowledge of the MEP pathway with emphasis on the understanding of the structure and the catalytic mechanism of the DXR enzyme and the global quest for therapeutically useful inhibitors of DXR.
Sigma-Aldrich
meso-Erythritol, ≥99% (GC)
Pricing and availability is not currently available.
Erythritol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.