Merck
  • A case of Ferguson-Smith type multiple keratoacanthomas associated with keratoacanthoma centrifugum marginatum: response to oral acitretin.
A case of Ferguson-Smith type multiple keratoacanthomas associated with keratoacanthoma centrifugum marginatum: response to oral acitretin.
Clinical and experimental dermatology (2007-08-24)
F Aydin, N Senturk, E Sabanciler, M T Canturk, A Y Turanli
ABSTRACT
Keratoacanthoma centrifugum marginatum (KCM) is a rare entity, usually classified as solitary keratoacanthoma (KA). The Ferguson-Smith type is the most common form of multiple KAs. Because development of multiple KAs and KCM in a single patient has rarely been reported, this association presents a therapeutic challenge. We report a 46-year-old man with Ferguson-Smith multiple KAs and KCM, who was successfully treated with acitretin.
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
USP
Acitretin, United States Pharmacopeia (USP) Reference Standard