Merck
  • Pharmacological, pharmacokinetic and clinical properties of olopatadine hydrochloride, a new antiallergic drug.
Pharmacological, pharmacokinetic and clinical properties of olopatadine hydrochloride, a new antiallergic drug.
Japanese journal of pharmacology (2002-06-06)
Kenji Ohmori, Ken-ichi Hayashi, Toshihiko Kaise, Etsuo Ohshima, Satoshi Kobayashi, Takashi Yamazaki, Akimichi Mukouyama
ABSTRACT
Olopatadine hydrochloride (olopatadine, 11-[(Z)-3-(dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid monohydrochloride) is a novel antiallergic/histamine H1-receptor antagonistic drug that was synthesized and evaluated in our laboratories. Oral administration of olopatadine at doses of 0.03 mg/kg or higher inhibited the symptoms of experimental allergic skin responses, rhinoconjunctivitis and bronchial asthma in sensitized guinea pigs and rats. Olopatadine is a selective histamine H1-receptor antagonist possessing inhibitory effects on the release of inflammatory lipid mediators such as leukotriene and thromboxane from human polymorphonuclear leukocytes and eosinophils. Olopatadine also inhibited the tachykininergic contraction in the guinea pig bronchi by prejunctional inhibition of peripheral sensory nerves. Olopatadine exerted no significant effects on action potential duration in isolated guinea pig ventricular myocytes, myocardium and human ether-a-go-go-related gene channel. Olopatadine was highly and rapidly absorbed in healthy human volunteers. The urinary excretion of olopatadine accounted for not less than 58% and the contribution of metabolism was considerably low in the clearance of olopatadine in humans. Olopatadine is one of the few renal clearance drugs in antiallergic drugs. Olopatadine was shown to be useful for the treatment of allergic rhinitis and chronic urticaria in double-blind clinical trials. Olopatadine was approved in Japan for the treatment of allergic rhinitis, chronic urticaria, eczema dermatitis, prurigo, pruritus cutaneous, psoriasis vulgaris and erythema exsudativum multiforme in December, 2000. Ophthalmic solution of olopatadine was also approved in the United States for the treatment of seasonal allergic conjunctivitis in December, 1996 (Appendix: also in the European Union, it was approved in February 2002).
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.
USP
Olopatadine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.