Merck
  • An unusual zwitterionic, diradical intermediate in photochemical electron-transfer substitution reactions.
An unusual zwitterionic, diradical intermediate in photochemical electron-transfer substitution reactions.
Journal of the American Chemical Society (2004-10-08)
John B Miller, Jeremy Breeding, Thomas J Smith
ABSTRACT
An unusual zwitterionic diradical intermediate complex R*...X-...HR'B*+ is generated during the photoinduced electron-transfer substitution reaction between alkyl halides (RX) and Lewis bases (HR'B). This reaction intermediate was observed for the 1-chloroadamantane and tert-butylamine system in a hydrocarbon glass at 25 K by electron spin resonance spectroscopy. Ab initio density functional calculations were carried out to examine the physical and electronic structure of the intermediate.
Sigma-Aldrich
1-Chloroadamantane, 98%