Merck
  • Orientational behavior of thermotropic liquid crystals on surfaces presenting electrostatically bound vesicular stomatitis virus.
Orientational behavior of thermotropic liquid crystals on surfaces presenting electrostatically bound vesicular stomatitis virus.
Langmuir : the ACS journal of surfaces and colloids (2005-04-20)
Luis A Tercero Espinoza, Kate R Schumann, Yan-Yeung Luk, Barbara A Israel, Nicholas L Abbott
ABSTRACT
We report the orientational behavior of nematic phases of 4-cyano-4'-pentylbiphenyl (5CB) on cationic, anionic, and nonionic surfaces before and after contact of these surfaces with solutions containing the negatively charged vesicular stomatitis virus (VSV). The surfaces were prepared on evaporated films of gold by either adsorption of poly-L-lysine (cationic) or formation of self-assembled monolayers (SAMs) from HS(CH2)2SO3- (anionic) or HS(CH2)11(OCH2CH2)4OH (nonionic). Prior to treatment with virus, we measured the initial orientation of 5CB (delta epsilon = epsilon(parallel) - epsilon(perpendicular) > 0) to be parallel to the cationic surfaces (planar anchoring) but perpendicular (homeotropic) after equilibration for 5 days. A similar transition from planar to homeotropic orientation of 5CB was observed on the anionic surfaces. Only planar orientations of 5CB were observed on the nonionic surfaces. Because N-(4-methoxybenzylidene)-4-butylaniline (MBBA, delta epsilon = epsilon(parallel) - epsilon(perpendicular) < 0) exhibited planar alignment on all surfaces, the time-dependent alignment of 5CB on the ionic surfaces is consistent with a dipolar coupling between the 5CB and electrical double layers formed at the ionic interfaces. Treatment ofpoly-L-lysine-coated gold films (cationic) with purified solutions of VSV containing 10(8)-10(10) plaque-forming units per milliliter (pfu/mL) led to the homeotropic alignment of 5CB immediately after contact of 5CB with the surface. In contrast, treatment of anionic surfaces and nonionic surfaces with solutions of VSV containing approximately 10(10) pfu/mL did not cause immediate homeotropic alignment of 5CB. These results and others suggest that homeotropic alignment of 5CB on cationic surfaces treated with VSV of titer > or = 10(8) pfu/mL reflects the presence of virus electrostatically bound to these surfaces.
Sigma-Aldrich
N-(4-Methoxybenzylidene)-4-butylaniline, 98%
Pricing and availability is not currently available.