Merck
  • Synthesis and chemical characterization of the novel agronomically relevant pentadentate chelate 2-(2-((2-hydroxybenzyl)amino)ethylamino)-2-(2-hydroxyphenyl)acetic acid (DCHA).
Synthesis and chemical characterization of the novel agronomically relevant pentadentate chelate 2-(2-((2-hydroxybenzyl)amino)ethylamino)-2-(2-hydroxyphenyl)acetic acid (DCHA).
Journal of agricultural and food chemistry (2010-06-10)
Sandra López-Rayo, Diana Hernández, Juan J Lucena, Rosa Escudero, Mar Gómez-Gallego, Miguel A Sierra
ABSTRACT
Iron chelates analogous to o,o-EDDHA/Fe(3+) are the fertilizers chosen to treat iron chlorosis in plants growing on calcareous soil. The isomer o,p-EDDHA/Fe(3+) presents less stability but faster assimilation by the plant than o,o-EDDHA/Fe(3+), because only five coordinating groups are able to complex Fe(3+). The new chelating agent 2-(2-((2-hydroxybenzyl)amino)ethylamino)-2-(2-hydroxyphenyl)acetic acid (DCHA) has been synthesized to obtain an iron fertilizer with intermediate stability between o,o-EDDHA/Fe(3+) and o,p-EDDHA/Fe(3+) and with fast assimilation. Its synthesis has been done starting from phenol, N-acetylethylendiamine, glyoxylic acid, and NaOH in a three-step sequence. The purity of the DCHA chelating agent, its protonation, and Ca(2+), Mg(2+), Fe(3+), and Cu(2+) stability constants, together with its ability to maintain iron in solution in different agronomic conditions, have been determined. The results indicate that the chelate DCHA/Fe(3+) has intermediate stability between those of o,o-EDDHA/Fe(3+) and o,p-EDDHA/Fe(3+) complexes and that it is capable of maintaining the Fe(3+) in agronomic conditions. This new chelating agent may be effective in correcting iron chlorosis in plants.
Sigma-Aldrich
Glyoxylic acid solution, 50 wt. % in H2O
Pricing and availability is not currently available.